Learn More
Sigma Laborzentrifugen™ 2-16KL Cooled Centrifuge with Angle Rotor
The refrigerated version of the Sigma 2-16P, this centrifuge is a compact all-rounder with excellent performance.
Brand: Sigma Laborzentrifugen™ 10350+12181
This item is not returnable.
View return policy
Description
- Universal laboratory centrifuge for swing-out and angle rotors
- Zero-maintenance brushless drive
- Selectable speed up to 15,300 rpm
- Rotor identification prevents overspeed operation
- Easy lid opening with gas spring support
- Window in lid for independent speed checking
- Spincontrol L
- Rotor cooling also possible at standstill; CFC-free R134a refrigerant
- Assured +4°C at maximum speed
Specifications
Angle rotor 12181 | |
550 mm | |
570 mm | |
720 |
-10°C to +40°C | |
310 mm | |
220/240 V | |
50/60 Hz |