Learn More
CD45 Mouse anti-Human, FITC, Clone: F10-89-4, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech FITC-65064-100TESTS
Beschreibung
The CD45 antibody from Proteintech is a mouse monoclonal antibody generated with purified t cells from human lymph nodes of human CD45. This antibody recognizes human antigen. The CD45 antibody has been validated for the following applications: FC analysis. This antibody is fitc.
Spezifikation
CD45 | |
Monoclonal | |
10 μL/Test | |
Mouse | |
Primary | |
Human |
Flow Cytometry | |
F10-89-4 | |
FITC | |
100 Tests | |
5788 | |
IgG2a |