Learn More
Cytohesin 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke:  Proteintech 10405-1-AP-20UL
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 16826773
  • 145.00 € / 20 Mikroliter
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
The protein encoded by this gene is a member of the PSCD family. Members of this family have identical structural organization that consists of an N-terminal coiled-coil motif, a central Sec7 domain, and a C-terminal pleckstrin homology (PH) domain. The coiled-coil motif is involved in homodimerization, the Sec7 domain contains guanine-nucleotide exchange protein (GEP) activity, and the PH domain interacts with phospholipids and is responsible for association of PSCDs with membranes. Members of this family appear to mediate the regulation of protein sorting and membrane trafficking. The encoded protein exhibits GEP activity in vitro with ARF1, ARF3, and ARF6 and is 83% homologous to CYTH1. Two transcript variants encoding different isoforms have been found for this gene.
Spezifikation
Spezifikation
Cytohesin 2
Polyclonal
Unconjugated
CYTH2
ARF exchange factor, ARNO, CTS18, CTS18.1, CYTH2, cytohesin 2, Protein ARNO, PSCD2, PSCD2L, SEC7L, Sec7p L, Sec7p like
Rabbit
Antigen Affinity Chromatography
RUO
116692, 19158, 9266
-20°C
Liquid
Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence
0.38 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P63034, P63035, Q99418
Cyth2
Cytohesin 2 Fusion Protein Ag0687
20 μL
Primary
Rat, Human, Mouse