Learn More
ECE2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 15418-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Converts big endothelin-1 to endothelin-1. Also involved in the processing of various neuroendocrine peptides, including neurotensin, angiotensin I, substance P, proenkephalin-derived peptides, and prodynorphin-derived peptides. May limit beta-amyloid peptide accumulation in brain. May also have methyltransferase activity.Spezifikation
ECE2 | |
Polyclonal | |
Unconjugated | |
ECE2 | |
ECE2, endothelin converting enzyme 2, KIAA0604 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
107522, 408243, 9718 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O60344, Q80Z60 | |
Ece2 | |
ECE2 Fusion Protein Ag7706 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |