Learn More
INTS7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 17054-1-AP-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This gene encodes a subunit of the integrator complex. The integrator complex associates with the C-terminal domain of RNA polymerase II and mediates 3′-end processing of the small nuclear RNAs U1 and U2. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.Spezifikation
INTS7 | |
Polyclonal | |
Unconjugated | |
INTS7 | |
C1orf73, INT7, integrator complex subunit 7, INTS7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
25896, 289382, 77065 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q7TQK1, Q9NVH2 | |
Ints7 | |
INTS7 Fusion Protein Ag10755 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |