Learn More
KISS1R Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 15505-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
GCPR54 is a galanin-like G protein coupled receptor that binds metastin, a peptide encoded by the metastasis suppressor gene KISS1. The tissue distribution of the protein suggests that it is involved in the regulation of endocrine function, and this is supported by the finding that it appears to play a role in the onset of puberty. GPR54 has been reported in various human brain regions, placenta, pancreas, and spinal cord, and at lower levels in spleen, peripheral blood leukocytes, testis, lymph node, thymus, small intestine, stomach, lung, and kidney. The receptor is also abundant in tumor tissues. In rat, GPR54 is expressed in liver, intestine, and in the pons, midbrain, thalamus, hypothalamus, hippocampus, amygdala, cortex (including frontal), and striatum. ESTs have been isolated from normal placenta and kidney cancer libraries.Spezifikation
KISS1R | |
Polyclonal | |
Unconjugated | |
KISS1R | |
AXOR12, G protein coupled receptor 54, GPR54, HOT7T175, Hypogonadotropin 1, KiSS 1 receptor, KiSS 1R, KISS1 receptor, KISS1R, Kisspeptins receptor, Metastin receptor | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
84634 | |
-20°C | |
Liquid |
Flow Cytometry, Western Blot, Immunofluorescence, Immunocytochemistry | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q969F8 | |
KISS1R | |
Peptide | |
20 μL | |
Primary | |
Human | |
Antibody | |