Learn More
Liebherr™ LFKV3953 Model Laboratory Refrigerator
Brand: Liebherr™ LFKV 3953
This item is not returnable.
View return policy
Specifications
Laboratory Refrigerator | |
4°C to 6°C | |
R600a | |
Glass Door | |
LFKV 3953 | |
43.5 cm | |
60 cm | |
150 V | |
150 w | |
150V | |
84 kg |
360 L | |
4 to 6°C | |
EC, ISO 14001, ISO 9001 | |
Automatic | |
61.5 cm | |
163.5 cm | |
44 cm | |
150 w | |
1.4kWh/24 hr. | |
90 kg |