Learn More
Systec™ Lifting Device for Loading/Unloading of Heavy Items for V-Series
For loading/unloading heavy items in V-Series autoclaves. Systec Lifting Device for Loading/Unloading of Heavy Items for V-Series is electrically operated with swivel arm and push button.
Brand:  Systec™ 9501
Product Code. 10610584
  • 5585.00€ / Each
Estimated Shipment: 15-07-2025
Explore more special offers
Add to basket
Description
Description
  • Electrically operated device with swivel arm facilitates loading and unloading of heavy items
  • Mobile control panel with push-button operation
  • Device is attached to side of autoclave
  • Swivel arm has special grip for baskets that remain stable even under loading conditions
  • Stainless steel wire mesh baskets, stackable
  • Stainless steel basket perforated only in upper third, sealed bottom for waste sterilization
  • Stainless steel basket perforated only in upper third, sealed bottom for waste sterilization with swivel lid
Specifications
Specifications
Loading/Unloading heavy items in V-series autoclaves
V-Series autoclaves
Lifting Device
Stainless Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers