Learn More
ANSELL™ Medium Weight Application EDGE™ Gloves
Good dexterity and flexibility and high cut protection and abrasion level. Good grip in dry environments with palm dipping for good protection against oil compared to a knitted or cut and sew glove.
Marke: ANSELL™ 48705050
78.32 EUR gültig bis 2025-06-30
Benutzen Sie den Promotions-Code "24156" um Ihren Promotions-Preis zu erhalten.
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- Seamless liner and knitted wrist offer close fit in cut-resistant applications.
- EDGE 48-705 offers high cut protection and abrasion resistance.
- Palm PU coating provides protection in dry and slightly oily environments.
- Cost-effective alternative compared to medium weight leather, thick cotton or aramid gloves.
Spezifikation
Ecovadis label|EN 388:2016 +A1:2018|EN ISO 21420:2020|CE Number|EN 420:2003 + A1:2009|ANSI Cut A3|ANSI ABR 6|Category II|REACH Compliant | |
Knit | |
Black | |
Spezifisch | |
Schnitt- und abriebfester Handschuh | |
4X43C | |
Unisex | |
Knit | |
Gerade | |
220 to 270 mm |
Palm and fingers | |
Polyurethan | |
ANSI A3 | |
Ja | |
5 | |
Aeronautics Assembly line Assembling metal parts and components Handling sharp or irregular metal pieces Molding Plastic injections | |
Black & Grey | |
Lined | |
13 |