Learn More
clikeco Neobox
Product-related services include: Setting up containers, collecting and replacing them, processing your waste through approved channels, managing your regulatory documents and issuing labels certifying your committed approach.
Brand: clikeco XXX24
This item is not returnable.
View return policy
Description
- Hermetically sealed for added safety
- Lightweight and compact, easy to move around
- Can be sealed to guarantee tamper-evidence
- Made of polypropylene for greater strength
- Stackable on lid for easy handling
- Easily identifiable thanks to clikeco colors
- Can be placed on the Themis for better visibility and sorting at source
- Complies with current European regulations
Product service includes:
- Analysis of your needs
- Provision of containers
- Collection and replacement of containers within 72 hours
- Transport to a local, approved processing center that recycles your waste.
- Issue and follow-up of your regulatory documents
- Delivery of the clikeco label and communication tools to promote your commitment
Specifications
UN certified | |
Soiled Solids Management | |
Neobox Tank | |
503.46 L | |
Leakproof Plastic Bag |
Blue, Orange | |
44 kg | |
133 gal. | |
Soiled Solid |