Learn More
PDLIM7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke:  Proteintech 17080-1-AP-150UL
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 16866914
  • 410.00 € / 150 Mikroliter
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Entdecken Sie weitere Sonderangebote
Beschreibung
Beschreibung
The protein encoded by this gene is representative of a family of proteins composed of conserved PDZ and LIM domains. LIM domains are proposed to function in protein-protein recognition in a variety of contexts including gene transcription and development and in cytoskeletal interaction. The LIM domains of this protein bind to protein kinases, whereas the PDZ domain binds to actin filaments. The gene product is involved in the assembly of an actin filament-associated complex essential for transmission of ret/ptc2 mitogenic signaling. The biological function is likely to be that of an adapter, with the PDZ domain localizing the LIM-binding proteins to actin filaments of both skeletal muscle and nonmuscle tissues. Alternative splicing of this gene results in multiple transcript variants.
Spezifikation
Spezifikation
PDLIM7
Polyclonal
Unconjugated
PDLIM7
ENIGMA, LIM mineralization protein, LMP, LMP1, PDLIM7, PDZ and LIM domain 7 (enigma), PDZ and LIM domain protein 7, Protein enigma
Rabbit
Antigen Affinity Chromatography
RUO
286908, 67399, 9260
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q3TJD7, Q9NR12, Q9Z1Z9
Pdlim7
PDLIM7 Fusion Protein Ag1334
150 μL
Primary
Human, Mouse, Rat