missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDLIM7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 17080-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The protein encoded by this gene is representative of a family of proteins composed of conserved PDZ and LIM domains. LIM domains are proposed to function in protein-protein recognition in a variety of contexts including gene transcription and development and in cytoskeletal interaction. The LIM domains of this protein bind to protein kinases, whereas the PDZ domain binds to actin filaments. The gene product is involved in the assembly of an actin filament-associated complex essential for transmission of ret/ptc2 mitogenic signaling. The biological function is likely to be that of an adapter, with the PDZ domain localizing the LIM-binding proteins to actin filaments of both skeletal muscle and nonmuscle tissues. Alternative splicing of this gene results in multiple transcript variants.Spezifikation
PDLIM7 | |
Polyclonal | |
Unconjugated | |
PDLIM7 | |
ENIGMA, LIM mineralization protein, LMP, LMP1, PDLIM7, PDZ and LIM domain 7 (enigma), PDZ and LIM domain protein 7, Protein enigma | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
286908, 67399, 9260 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3TJD7, Q9NR12, Q9Z1Z9 | |
Pdlim7 | |
PDLIM7 Fusion Protein Ag1334 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur