Learn More
Pepsinogen II Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 16881-1-AP-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Hydrolyzes a variety of proteins.Spezifikation
Pepsinogen II | |
Polyclonal | |
Unconjugated | |
PGC | |
Gastricsin, Pepsinogen C, Pepsinogen II, PEPSINOGEN, GROUP II, PGC, Progastricsin, progastricsin (pepsinogen C) | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
109820, 24864, 5225 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P04073, P20142, Q9D7R7 | |
PGC | |
Pepsinogen II Fusion Protein Ag10332 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |