Learn More
PTEN Mouse anti-Human, CoraLite 488, Clone: 5C10B6, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech CL488-60300-100UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The PTEN antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PTEN. This antibody recognizes human, mouse antigen. The PTEN antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Spezifikation
PTEN | |
Monoclonal | |
2000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse |
Immunofluorescence | |
5C10B6 | |
CoraLite 488 | |
100 μL | |
5728 | |
IgG2a |