Learn More
Sartorius™ Calibration Express Mail-In Pipette Calibration Services
Shipping service provides high-quality pipette calibrations with simplified service options, transit boxes and forms combined with 48-hour turnaround time
Specifications
Certifications/Compliance | ISO 8655 and ISO 17025 |
---|
Includes: Calibration Express boxes come with instructions and service forms inside.
Description
- Computer generated Calibration Certificates
- Each pipette receives sticker with date of service and next calibration due date
- Calibrated to manufacturer's specifications
- As Found calibration data
- Three levels of service (Basic, Premier and Elite) to meet lab's calibration needs
- Secure transit boxes and packing material with each shipment
- Round trip transportation by FedEx ground service
- Simplified service request and decontamination forms
- All service tiers offer single channel and multi-channel (8 and 12 channels)
- Upgrade to Complete maintenance care options of Cleaning and Lube - save on long term repairs
- All come with As Found Data, Calibration Certificate and Sticker
- Second day and overnight delivery option are available
Basic
- 4.3 Calibration Service Includes: four readings at three volumes (100%, 50%, 10% of nominal volume)
Premier
- 5.3 Calibration Service Includes: five readings at three volumes (100%,50%, 10% of nominal volumes)
Elite
- 10.3 Calibration Service Includes: ISO 17025/8655 compliant ten readings at three volumes (100%, 50%, 10% of nominal)
All manufacturers' pipettes serviced. Return Ground Shipping via FedEx is included.
Certifications
ISO 8655 and ISO 17025
Specifications
ISO 8655 and ISO 17025 |
Customer must pay shipping costs to send pipettes to Sartorius for calibration.