Learn More
Copan Italia™ Stuart Agar Gel Swab
Single plastic swab suitable for throat, vaginal, wound, and skin swabs. Copan Italia™ Stuart Agar Gel Swab is coated with a non-nutrient soft agar gel, and designed for specimen transportation.
Brand: Copan Italia™ 111C.USE
This item is not returnable.
View return policy
Description
- Single plastic swab
- Stuart Agar gel medium
- Suitable for throat, vaginal, wound, and skin swabs
- For specimen transportation
Specifications
Stuart Agar | |
Throat, vaginal, wound, and skin swabs | |
50/Pk., 10 x 50/Cs. |
Blue | |
Plastic |