Learn More
TNFAIP3 Mouse anti-Human, CoraLite 488, Clone: 3A11G6, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech CL488-66695-100UL
Beschreibung
The TNFAIP3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TNFAIP3. This antibody recognizes human, rat antigen. The TNFAIP3 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Spezifikation
TNFAIP3 | |
Monoclonal | |
2000 μg/mL | |
Mouse | |
Primary | |
Human, Rat |
Immunofluorescence | |
3A11G6 | |
CoraLite 488 | |
100 μL | |
7128 | |
IgG1 |