Learn More
Calpain 3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke:  Proteintech 10415-1-AP-20UL
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 16826793
  • 145.00 € / 20 Mikroliter
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Calpain, a heterodimer consisting of a large and a small subunit, is a major intracellular protease, although its function has not been well established. This gene encodes a muscle-specific member of the calpain large subunit family that specifically binds to titin. Mutations in this gene are associated with limb-girdle muscular dystrophies type 2A. Alternate promoters and alternative splicing result in multiple transcript variants encoding different isoforms and some variants are ubiquitously expressed.
Spezifikation
Spezifikation
Calpain 3
Polyclonal
Unconjugated
CAPN3
Calpain 3, calpain 3, (p94), Calpain L3, Calpain p94, CANP 3, CANP3, CANPL3, CAPN3, LGMD2, LGMD2A, nCL 1, NCL1, New calpain 1, p94
Rabbit
Antigen Affinity Chromatography
RUO
12335, 29155, 825
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P16259, P20807, Q64691
Capn3
Calpain 3 Fusion Protein Ag0671
20 μL
Primary
Mouse, Human, Rat