Learn More
DMRTC1B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 21473-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
DMRTC1B, also named as Doublesex- and mab-3-related transcription factor C1, is a 192 amino acid protein, which belongs to the DMRT family. DMRTC1B is predomitly expressed in kidney, pancreas, ovary and testis. Detected in brain and in many other tissues. DMRTC1B exists two isoform and molecular weight of isoforms are 20 kDa and 13 kDa.Spezifikation
DMRTC1B | |
Polyclonal | |
Unconjugated | |
DMRTC1B | |
DMRT like family C1B, DMRTC1, DMRTC1B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
632708, 728656 | |
-20°C | |
Liquid |
Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5HYR2 | |
Dmrtc1b | |
DMRTC1B Fusion Protein Ag15419 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |