Learn More
GAT1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20298-1-AP-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The GAT1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human GAT1. This antibody recognizes human, mouse , rat antigen. The GAT1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spezifikation
GAT1 | |
Polyclonal | |
Unconjugated | |
150 μL | |
6529 | |
IgG |
Western Blot, Immunohistochemistry, ELISA | |
333 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |