Learn More
Glucocorticoid receptor Mouse anti-Human, Unconjugated, Clone: 1D9A9, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66904-1-IG-20UL
Beschreibung
The Glucocorticoid receptor antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Glucocorticoid receptor. This antibody recognizes human, mouse, rat antigen. The Glucocorticoid receptor antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Spezifikation
Glucocorticoid receptor | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
1D9A9 | |
Unconjugated | |
20 μL | |
2908 | |
IgG1 |