Learn More
NR4A3 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 55405-1-AP-150UL
 View more versions of this product
Product Code. 16866865
  • 410.00 € / 150µL
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
Description
Description
The NR4A3 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human NR4A3. This antibody recognizes human, mouse antigen. The NR4A3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
NR4A3
Polyclonal
Unconjugated
150 μL
8013
IgG
Western Blot, ELISA
260 μg/mL
Rabbit
Primary
Human, Mouse